Learn More
Sigma Aldrich Fine Chemicals Biosciences fibroblast Growth factor B
Catalog No. 502904755
Encompass
Since fGf-Basic is found in a variety of organs acts on a wide range of cell types and has multifunctional actions it has acquired numerous synonyms including heparin-binding growth factor (class II or beta) eye-derived growth factor I cartilage-derived growth factor and astroglial growth factor II5. Purified bovine and human fGf-basic differ by 3 amino acids in sequence3 and are biologically and immunologically cross-reactive.
Catalog No. 50-290-4755 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F368525UG
Since fGf-Basic is found in a variety of organs acts on a wide range of cell types and has multifunctional actions it has acquired numerous synonyms including heparin-binding growth factor (class II or beta) eye-derived growth factor I cartilage-derived growth factor and astroglial growth factor II5. Purified bovine and human fGf-basic differ by 3 amino acids in sequence3 and are biologically and immunologically cross-reactive.