Learn More
Cayman Chemical Fibroblast Growth Factor Receptors 1 & 2 Assay for Paracrine FGF Signaling (FGFR1/2), 3x32 Well
SDP
Catalog No. 502910453
Encompass_Preferred
Reporter Cells are transiently transfected and prepared as frozen stocks using INDIGO's proprietary CryoMite™ process. This cryo-preservation method allows for the immediate dispensing of healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup.
Catalog No. 50-291-0453 Supplier Cayman Chemical Supplier No. 347853X32 WELL
Reporter Cells are transiently transfected and prepared as frozen stocks using INDIGO's proprietary CryoMite™ process. This cryo-preservation method allows for the immediate dispensing of healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup.