Learn More
ATCC Fibroblast Growth Kit-Low serum
Catalog No. 502382961
Encompass
Fibroblast Growth Kit–Serum-Free (ATCC PCS-201-040) and Fibroblast Growth Kit–Low Serum (ATCC PCS-201-041) each contain components that when added to Fibroblast Basal Medium (ATCC PCS-201-030) create a complete ATCC Primary Cell Solutions™ culture environment for fibroblasts (e.g., Dermal Fibroblasts, Normal, Human Neonatal [ATCC PCS-201-010], Dermal Fibroblasts, Normal, Human Neonatal, Mitomycin C Treated [ATCC PCS-201-010]).
Catalog No. 50-238-2961 Supplier ATCC Supplier No. PCS201041
Fibroblast Growth Kit–Serum-Free (ATCC PCS-201-040) and Fibroblast Growth Kit–Low Serum (ATCC PCS-201-041) each contain components that when added to Fibroblast Basal Medium (ATCC PCS-201-030) create a complete ATCC Primary Cell Solutions™ culture environment for fibroblasts (e.g., Dermal Fibroblasts, Normal, Human Neonatal [ATCC PCS-201-010], Dermal Fibroblasts, Normal, Human Neonatal, Mitomycin C Treated [ATCC PCS-201-010]).