Learn More
ATCC Fibroblast Growth Kit-Serum-free
Catalog No. 502382071
Encompass
Fibroblast Growth Kit–Serum-free (ATCC PCS-201-040) and Fibroblast Growth Kit–Low Serum (ATCC PCS-201-041) each contain components that when added to Fibroblast Basal Medium (ATCC PCS-201-030) creates a complete ATCC Primary Cell Solutions culture environment for fibroblasts (eg, Dermal Fibroblasts, Normal, Human Neonatal, ATCC PCS-201-010).
Catalog No. 50-238-2071 Supplier ATCC Supplier No. PCS201040
Fibroblast Growth Kit–Serum-free (ATCC PCS-201-040) and Fibroblast Growth Kit–Low Serum (ATCC PCS-201-041) each contain components that when added to Fibroblast Basal Medium (ATCC PCS-201-030) creates a complete ATCC Primary Cell Solutions culture environment for fibroblasts (eg, Dermal Fibroblasts, Normal, Human Neonatal, ATCC PCS-201-010).