Learn More
Cayman Chemical Fibroblast Growth Factor Receptor 1c and β-Klotho (FGFR/β-Klotho) for Endocrine FGF Signaling Reporter Assay System, 384 Well
SDP
Catalog No. 502910510
Encompass_Preferred
FGFR/β-Klotho Reporter Cells are transiently transfected and prepared as frozen stocks using INDIGO's proprietary CryoMite™ process. This cryo-preservation method allows for the immediate dispensing of healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup.
Catalog No. 50-291-0510 Supplier Cayman Chemical Supplier No. 34959384 WELL
FGFR/β-Klotho Reporter Cells are transiently transfected and prepared as frozen stocks using INDIGO's proprietary CryoMite™ process. This cryo-preservation method allows for the immediate dispensing of healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup.