Learn More
Cayman Chemical Fibroblast Growth Factor Receptor 4 (FGFR) Reporter Assay System, 96 Well
SDP
Catalog No. 502943556
Encompass_Preferred
This assay kit provides the convenience of an all-inclusive cell-based reporter assay system for FGFR4 paracrine FGF signaling. In addition to Reporter Cells, included are an optimized culture medium for use in reviving the cryopreserved cells, a culture medium supplemented with heparin for use in preparing test sample treatments, the physiological paracrine activator FGF-2 (FGF-Basic), Luciferase Detection Reagents, and a cell culture-ready assay plate.
Catalog No. 50-294-3556 Supplier Cayman Chemical Supplier No. 4225496 WELL
This assay kit provides the convenience of an all-inclusive cell-based reporter assay system for FGFR4 paracrine FGF signaling. In addition to Reporter Cells, included are an optimized culture medium for use in reviving the cryopreserved cells, a culture medium supplemented with heparin for use in preparing test sample treatments, the physiological paracrine activator FGF-2 (FGF-Basic), Luciferase Detection Reagents, and a cell culture-ready assay plate.