Learn More
Sciencell Research Laboratories FIBROBLAST MEDIUM-2
Catalog No. NC0665133
Encompass
Fm-2 Consists Of 500 Ml Of Basal Medium, 25 Ml Of Fetal Bovine Serum (Fbs, Cat. No. 0025), 5 Ml Of Fibroblast Growth Supplement-2 (Fgs-2, Cat. No. 2382), And 5 Ml Of Penicillin/Streptomycin Solution (P/S, Cat. No. 0503).
Catalog No. NC0665133 Supplier Sciencell Research Laboratories Supplier No. 2331
Fm-2 Consists Of 500 Ml Of Basal Medium, 25 Ml Of Fetal Bovine Serum (Fbs, Cat. No. 0025), 5 Ml Of Fibroblast Growth Supplement-2 (Fgs-2, Cat. No. 2382), And 5 Ml Of Penicillin/Streptomycin Solution (P/S, Cat. No. 0503).