Learn More
Sciencell Research Laboratories FIBROBLAST MEDIUM 500ML
Catalog No. NC9772540
Encompass
Fibroblast medium is a complete medium designed for optimal growth of normal human fibroblasts in vitro. It is shipped as a kit with FBS, fibroblast growth supplement, and Pen/Strep solution.
Catalog No. NC9772540 Supplier Sciencell Research Laboratories Supplier No. 2301
Fibroblast medium is a complete medium designed for optimal growth of normal human fibroblasts in vitro. It is shipped as a kit with FBS, fibroblast growth supplement, and Pen/Strep solution.