Learn More
Lifeline Cell Technology Fibroblast Supp. Kit
SDP
Catalog No. NC1944660
Encompass_Preferred
FibroLife S2 LifeFactors Kit. Supplements for culturing fibroblasts in a 2% FBS environment. Best paired with LM-0001 FibroLife Basal Medium.
Catalog No. NC1944660 Supplier Lifeline Cell Technology Supplier No. LS1038
FibroLife S2 LifeFactors Kit. Supplements for culturing fibroblasts in a 2% FBS environment. Best paired with LM-0001 FibroLife Basal Medium.