Learn More
Promocell GmbH PromoCell Fibroblast Growth Medium 2 KIT 500 ml, Contains: C-23220, C-39320
Catalog No. 50306218
Encompass
PromoCell Fibroblast Growth Medium 2 is a low-serum (2% V/V) medium and has been developed for the in vitro cultivation of adult Human Fibroblasts.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for bovine, porcine, and rat fibroblasts, as well as fibroblast cell lines.| Media supplement | Final supplement concentration|| Fetal Calf Serum | 0.02 ml / ml || Basic Fibroblast Growth Factor (recombinant human) | 1 ng / ml || Insulin (recombinant human) | 5 μg / ml |
Catalog No. 50-306-218 Supplier Promocell GmbH Supplier No. C23120
PromoCell Fibroblast Growth Medium 2 is a low-serum (2% V/V) medium and has been developed for the in vitro cultivation of adult Human Fibroblasts.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for bovine, porcine, and rat fibroblasts, as well as fibroblast cell lines.| Media supplement | Final supplement concentration|| Fetal Calf Serum | 0.02 ml / ml || Basic Fibroblast Growth Factor (recombinant human) | 1 ng / ml || Insulin (recombinant human) | 5 μg / ml |