Learn More
Diagnostic Biosystems FIBRONECTIN Fibronectin
Catalog No. 502407476
Encompass_Preferred
This antibody reacts with human fibronectin in liver, tonsil, skin and kidney. Traces of contaminating antibodies have been removed by solid-phase adsorption. Only the fibronectin arch was observed in crossed immunoelectrophoresis.
Catalog No. 50-240-7476 Supplier Diagnostic Biosystems Supplier No. PDR170
This antibody reacts with human fibronectin in liver, tonsil, skin and kidney. Traces of contaminating antibodies have been removed by solid-phase adsorption. Only the fibronectin arch was observed in crossed immunoelectrophoresis.