Learn More
Santa Cruz Biotechnology FIBRONECTIN ANTI EP5 200UG/ML
Supplier:  Santa Cruz Biotechnology SC8422
Encompass
Fibronectin Antibody (EP5) is a mouse monoclonal IgG1 (kappa light chain) provided at 200 ug/ml
Catalog No. NC1714192