Learn More
CPC Scientific H-Phe-Asn-Lys-His-Thr-Glu-Ile-Ile-Glu-Glu-Asp-Thr-Asn-Lys-Asp-Lys-Pro-Ser-Tyr-Gln-Phe-Gly-Gly-His-Asn-Ser-Val-Asp-Phe-Glu-Glu-Asp-Thr-Leu-Pro-Lys-Val-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific FIBN008A
Encompass
A synthetic peptide that mimics the structure of a 38-amino acid unit from a staphylococcal fibronectin-binding protein. This peptide represents the protein domain to which the fibronectin-binding activity has been localized and also inhibits the binding of fibronectin to bacterial cells.SEQUENCE: H-Phe-Asn-Lys-His-Thr-Glu-Ile-Ile-Glu-Glu-Asp-Thr-Asn-Lys-Asp-Lys-Pro-Ser-Tyr-Gln-Phe-Gly-Gly-His-Asn-Ser-Val-Asp-Phe-Glu-Glu-Asp-Thr-Leu-Pro-Lys-Val-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: FNKHTEIIEEDTNKDKPSYQFGGHNSVDFEEDTLPKVMOLECULAR FORMULA: C190H283N49O66MOLECULAR WEIGHT: 4309.7STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [119977-20-7]SYNONYMS: RESEARCH AREA: InflammationREFERENCES:C. Signas et al., PNAS, 86, 699 (1989)SKU(s): FIBN-008A, FIBN-008B
Catalog No. 50-210-9283