Learn More
Cytoskeleton, Inc. Fibronectin (Biotinylated), High Purity Bovine Plasma Source, Lyophilized Powder, 20 µg
Supplier:  Cytoskeleton, Inc. FNR03A
Encompass
Fibronectin (Biotinylated) from Cytoskeleton, Inc. is a high-purity, bovine plasma-derived glycoprotein that enables optimal cell adhesion, matrix assembly, and migration research. This lyophilized powder boasts over 80% purity and features a unique biotinylation method enhancing its detection capabilities. Ideal for applications in cell invasion assays and bioengineering, it provides researchers with a stable, convenient product for investigating cell-polymer interactions.
Catalog No. NC0827107