Learn More
Sigma Aldrich Fine Chemicals Biosciences fibronectin bovine plasma2MG
Catalog No. 502904893
Encompass
fibronectin is a glycoprotein with multiple domains. It comprises a N-terminal central binding domain and a heparin-binding domain. Bovine plasma fibronectin has polypeptide chains with disulfide linkage and corresponds to a molecular weight of 220 and 215 kDa.
Catalog No. 50-290-4893 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F47592MG
fibronectin is a glycoprotein with multiple domains. It comprises a N-terminal central binding domain and a heparin-binding domain. Bovine plasma fibronectin has polypeptide chains with disulfide linkage and corresponds to a molecular weight of 220 and 215 kDa.