Learn More
Sigma Aldrich Fine Chemicals Biosciences fibronectin bovine plasma5MG
Catalog No. 502914179
Encompass
fibronectin is a glycoprotein with multiple domains. It comprises a N-terminal central binding domain and a heparin-binding domain. Bovine plasma fibronectin has polypeptide chains with disulfide linkage and corresponds to a molecular weight of 220 and 215 kDa.
Catalog No. 50-291-4179 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F47595MG
fibronectin is a glycoprotein with multiple domains. It comprises a N-terminal central binding domain and a heparin-binding domain. Bovine plasma fibronectin has polypeptide chains with disulfide linkage and corresponds to a molecular weight of 220 and 215 kDa.