Learn More
Bioss Fibronectin Cell/Heparin2 Bind
SDP
Catalog No. 5000065798
Encompass_Preferred
Fibronectin (Cell/Heparin2 Binding region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications WB IP M013Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-65798 Supplier Bioss Supplier No. BSM70501M
Fibronectin (Cell/Heparin2 Binding region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications WB IP M013Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol