Learn More
Cusabio Technology LLC fibronectin FN autoantibody
Catalog No. 5000049780
Encompass_Preferred
The Human Fibronectin (FN) Autoantibody ELISA Kit is designed to detect FN auto-Ab in human serum and plasma samples using an indirect qualitative method This assay provides results within 1 to 5 hours requiring only 50-100 L of sample per test The kit utilizes a 450 nm wavelength for absorbance measurement ensuring specific detection of autoantibodies against fibronectin It is optimized for research and diagnostic applications to identify the presence of FN auto-Ab aiding in studies related to autoimmune disorders tissue remodeling and inflammatory conditions The indirect ELISA format ensures high sensitivity by employing a secondary enzyme-linked antibody for signal amplification With standardized protocols and reliable performance this kit serves as a practical tool for laboratories focused on autoantibody profiling and immunological research
Catalog No. 50-000-49780 Supplier Cusabio Technology LLC Supplier No. CSBEQ027225HU
The Human Fibronectin (FN) Autoantibody ELISA Kit is designed to detect FN auto-Ab in human serum and plasma samples using an indirect qualitative method This assay provides results within 1 to 5 hours requiring only 50-100 L of sample per test The kit utilizes a 450 nm wavelength for absorbance measurement ensuring specific detection of autoantibodies against fibronectin It is optimized for research and diagnostic applications to identify the presence of FN auto-Ab aiding in studies related to autoimmune disorders tissue remodeling and inflammatory conditions The indirect ELISA format ensures high sensitivity by employing a secondary enzyme-linked antibody for signal amplification With standardized protocols and reliable performance this kit serves as a practical tool for laboratories focused on autoantibody profiling and immunological research