Learn More
Sigma Aldrich Fine Chemicals Biosciences fibronectin from human pla
Catalog No. 502905729
Encompass
fibronectin is responsible for the cellular interactions with the extracellular matrix. It is involved in cell adhesion migration growth and differentiation. It is a soluble component of plasma and other body fluids. In addition it is also part of the insoluble extracellular matrix.
Catalog No. 50-290-5729 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10838039001
fibronectin is responsible for the cellular interactions with the extracellular matrix. It is involved in cell adhesion migration growth and differentiation. It is a soluble component of plasma and other body fluids. In addition it is also part of the insoluble extracellular matrix.