Learn More
Sigma Aldrich Fine Chemicals Biosciences fibronectin human plasma l2MG
Catalog No. 502904611
Encompass
fibronectin 1 is a glycoprotein of the extracellular matrix that is coded by fN1 gene. It is expressed in the plasma and at the cell surface. The fN1 gene mapped to human chromosome 2q35.
Catalog No. 50-290-4611 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F08952MG
fibronectin 1 is a glycoprotein of the extracellular matrix that is coded by fN1 gene. It is expressed in the plasma and at the cell surface. The fN1 gene mapped to human chromosome 2q35.