Learn More
Abcam Mouse Fibronectin ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1088491X96TESTS
Encompass_Preferred
Mouse Fibronectin ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Fibronectin in Mouse in Urine, Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0971084