Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibronectin, MP Biomedicals™

Catalog No. MP215112601 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215112601 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215112601 Supplier MP Biomedicals, Inc Supplier No. 0215112601
Add to Cart
Add to Cart

Found both as cell surface proteins and in plasma. Fibronectin, MP Biomedicals is an adhesive glycoprotein suitable for use as an attachment factor in the propagation of cells in vitro.

Human fibronectin (HFN) is suitable for use as an attachment factor in the propagation of cells in vitro when used to coat cell culture surfaces, including plasticware, glassware, and microcarrier beads. It is used in cell attachment and proliferation assays on human endothelial cells, human keratinocytes and human dermal fibroblasts.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86088-83-7
Synonym CIG, Cold insoluble globulin, Fibronectin from human plasma=

Specifications

Quantity 1 mg
Solubility Information Soluble in water (1mg/mL at 37°C).
Percent Purity ≥95%
Physical Form Solid
Recommended Storage Store at −20°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.