Learn More
Sigma Aldrich Fine Chemicals Biosciences fibronectin pure
Catalog No. 502914426
Encompass
fibronectin is responsible for the cellular interactions with the extracellular matrix. It is involved in cell adhesion migration growth and differentiation. It is a soluble component of plasma and other body fluids. In addition it is also part of the insoluble extracellular matrix.
Catalog No. 50-291-4426 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11080938001
fibronectin is responsible for the cellular interactions with the extracellular matrix. It is involved in cell adhesion migration growth and differentiation. It is a soluble component of plasma and other body fluids. In addition it is also part of the insoluble extracellular matrix.