Learn More
Sigma Aldrich Fine Chemicals Biosciences fibronectin rat plasma pow5MG
Catalog No. 502904490
Encompass
fibronectin (fN)/fN1 (fibronectin 1) is a multifunctional glycoprotein produced by hepatic stellate cells (HSCs). It is a constituent of extracellular matrix (ECM). fN1 is found in the cell membrane and cytoplasm. It is also synthesized and liberated by fibroblasts endothelial cells macrophages and hepatocytes in vivo.
Catalog No. 50-290-4490 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F0635.5MG
fibronectin (fN)/fN1 (fibronectin 1) is a multifunctional glycoprotein produced by hepatic stellate cells (HSCs). It is a constituent of extracellular matrix (ECM). fN1 is found in the cell membrane and cytoplasm. It is also synthesized and liberated by fibroblasts endothelial cells macrophages and hepatocytes in vivo.