Learn More
Aviva Systems Biology Corp FICD Antibody - C-terminal region (ARP46825_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46825_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against FICD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human FICD. Alternate names: HYPE, MGC5623, UNQ3041, hip13, HIP13ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-53493