Learn More
Thermo Scientific Chemicals Ficin, fig tree latex
Catalog No. AAJ6013614
Change view
Click to view available options
Quantity:
25 g
Catalog No. Quantity
AAJ6013614 25 g
1 options
Catalog No. AAJ6013614 Supplier Thermo Scientific Chemicals Supplier No. J6013614
Add to Cart
Immobilized Ficin was specifically designed for cleavage of mouse IgG1 into F(ab')2 or Fab fragments. Ficin acts on bonds involving uncharged and/or aromatic amino acids. The immobilization of ficin enhances stability against denaturation, heat and autolysis. Immobilization also eliminates any potential for antibody-enzyme adducts that cause continued sample digestion. In addition to these advantages, immobilization allows the ficin to be reused.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25 g
Packaging Glass bottle
Form Lyophilized