Learn More
Research Products International Corp Ficoll 400 [Polysucrose 400], 5 Grams
SDP
Catalog No. 50488658
Encompass_Preferred
Ficoll is an uncharged polymer of sucrose widely used for stabilizing cells and isolation of nuclei and organelles. Also used in Denhardt's Reagent and in gel loading buffers.
Catalog No. 50-488-658 Supplier Research Products International Corp Supplier No. F104005.0
Ficoll is an uncharged polymer of sucrose widely used for stabilizing cells and isolation of nuclei and organelles. Also used in Denhardt's Reagent and in gel loading buffers.