Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ficoll(R) 400, BiotechGrade, Spectrum™ Chemical
SDP

Catalog No. 18602727 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-727 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-602-727 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F30085GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

F3008, 26873-85-8

Spectrum™ Chemical Ficoll(R) 400, BiotechGrade

Chemical Identifiers

CAS 26873-85-8
Molecular Formula C15H27ClO12
Molecular Weight (g/mol) 434.82
InChI Key IACFXVUNKCXYJM-UHFFFAOYNA-N
IUPAC Name 2-(chloromethyl)oxirane, 2-{[3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES ClCC1CO1.OCC1OC(CO)(OC2OC(CO)C(O)C(O)C2O)C(O)C1O

Specifications

CAS Max % 100%
Quantity 5 g
Packaging Amber Glass Bottle
Grade Biotech
Chemical Name or Material Ficoll(R) 400
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.