Learn More
Crescent Chemical Co Inc FICOLL 401
SDP
Catalog No. 50247591
Encompass_Preferred
Ficoll 400 Suited for preparation of continuous and discontinuous density gradients. For stabilization of membrane-bound particles, isolation of lymphocytes. Dialyzable material (including NaCl) max. 1 %. Better osmotic properties than sucrose, it preserves
Catalog No. 50-247-591 Supplier Crescent Chemical Co Inc Supplier No. SE21373.02
Ficoll 400 Suited for preparation of continuous and discontinuous density gradients. For stabilization of membrane-bound particles, isolation of lymphocytes. Dialyzable material (including NaCl) max. 1 %. Better osmotic properties than sucrose, it preserves