Learn More
Sigma Aldrich Fine Chemicals Biosciences ficollR 400 BioXtra for mo100G
Catalog No. 502902476
Encompass
ficoll(R) 400 is a non-ionic synthetic polymer of sucrose used for cell separation and organ isolation.
Catalog No. 50-290-2476 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F2637100G
ficoll(R) 400 is a non-ionic synthetic polymer of sucrose used for cell separation and organ isolation.