Learn More
Sigma Aldrich Fine Chemicals Biosciences FicollR Paque Plus Cytiva
Catalog No. 502908436
Encompass
Ficoll-Paque Plus is a recognized standard in laboratories worldwide for the isolation of human lymphocytes for in vitro studies.Ficoll-Paque PLUS is a ready to use sterile medium for isolation of lymphocytes in high yield from peripheral blood using a simple and rapid centrifugation procedure. It maintains the viability and a representative distribution of B and T lymphocytes.
Catalog No. 50-290-8436 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17144003
Ficoll-Paque Plus is a recognized standard in laboratories worldwide for the isolation of human lymphocytes for in vitro studies.Ficoll-Paque PLUS is a ready to use sterile medium for isolation of lymphocytes in high yield from peripheral blood using a simple and rapid centrifugation procedure. It maintains the viability and a representative distribution of B and T lymphocytes.