Learn More
Sigma Aldrich Fine Chemicals Biosciences ficollR Paque Plus Cytiva
Catalog No. 502904331
Encompass
ficoll-Paque Plus is a recognized standard in laboratories worldwide for the isolation of human lymphocytes for in vitro studies.ficoll-Paque PLUS is a ready to use sterile medium for isolation of lymphocytes in high yield from peripheral blood using a simple and rapid centrifugation procedure. It maintains the viability and a representative distribution of B and T lymphocytes.
Catalog No. 50-290-4331 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17144002
ficoll-Paque Plus is a recognized standard in laboratories worldwide for the isolation of human lymphocytes for in vitro studies.ficoll-Paque PLUS is a ready to use sterile medium for isolation of lymphocytes in high yield from peripheral blood using a simple and rapid centrifugation procedure. It maintains the viability and a representative distribution of B and T lymphocytes.