Learn More
Sigma Aldrich Fine Chemicals Biosciences ficollR Paque PREMIUM 1.07
Catalog No. 502904216
Encompass
ficoll-Paque PREMIUM products are a range of sterile ready-to-use density gradient media for the preparation of mononuclear cells for use in cell therapy applications.
Catalog No. 50-290-4216 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17544652
ficoll-Paque PREMIUM products are a range of sterile ready-to-use density gradient media for the preparation of mononuclear cells for use in cell therapy applications.