Learn More
Sigma Aldrich Fine Chemicals Biosciences FicollR Paque Premium Cyti
Catalog No. 502907531
Encompass
Ficoll-Paque PREMIUM products are a range of sterile ready-to-use density gradient media for the preparation of mononuclear cells.Ficoll-Paque PREMIUM products are based on Ficoll-Paque PLUS (see Ficoll-Paque PLUS) which has been used for more than 30 yr as a sterile density medium for the isolation of high yields of mononuclear cells from bone marrow peripheral blood and cord blood. All Ficoll-Paque PREMIUM products differ from Ficoll-Paque PLUS in that they are manufactured under a Quality Management System certified to ISO 134852003.
Catalog No. 50-290-7531 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17544203
Ficoll-Paque PREMIUM products are a range of sterile ready-to-use density gradient media for the preparation of mononuclear cells.Ficoll-Paque PREMIUM products are based on Ficoll-Paque PLUS (see Ficoll-Paque PLUS) which has been used for more than 30 yr as a sterile density medium for the isolation of high yields of mononuclear cells from bone marrow peripheral blood and cord blood. All Ficoll-Paque PREMIUM products differ from Ficoll-Paque PLUS in that they are manufactured under a Quality Management System certified to ISO 134852003.