Learn More
Sigma Aldrich Fine Chemicals Biosciences ficollR-Paque Premium Cyti
Catalog No. 502904660
Encompass
ficoll-Paque PREMIUM products are a range of sterile ready-to-use density gradient media for the preparation of mononuclear cells.ficoll-Paque PREMIUM products are based on ficoll-Paque PLUS (see ficoll-Paque PLUS) which has been used for more than 30 yr as a sterile density medium for the isolation of high yields of mononuclear cells from bone marrow peripheral blood and cord blood. All ficoll-Paque PREMIUM products differ from ficoll-Paque PLUS in that they are manufactured under a Quality Management System certified to ISO 134852003.
Catalog No. 50-290-4660 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17544202
ficoll-Paque PREMIUM products are a range of sterile ready-to-use density gradient media for the preparation of mononuclear cells.ficoll-Paque PREMIUM products are based on ficoll-Paque PLUS (see ficoll-Paque PLUS) which has been used for more than 30 yr as a sterile density medium for the isolation of high yields of mononuclear cells from bone marrow peripheral blood and cord blood. All ficoll-Paque PREMIUM products differ from ficoll-Paque PLUS in that they are manufactured under a Quality Management System certified to ISO 134852003.