Learn More
Sigma Aldrich Fine Chemicals Biosciences ficollR PM 70 Cytiva 17 03
Catalog No. 502904867
Encompass
High molecular weight sucrose-polymers formed by copolymerization of sucrose with epichlorohydrin. The molecules are highly branched and the high content of hydroxyl groups leads to very good solubility in aqueous media.
Catalog No. 50-290-4867 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17031010
High molecular weight sucrose-polymers formed by copolymerization of sucrose with epichlorohydrin. The molecules are highly branched and the high content of hydroxyl groups leads to very good solubility in aqueous media.