Learn More
Medchemexpress LLC HY-12451 10mg , FICZ CAS:172922-91-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1245110MG
Encompass_Preferred
HY-12451 10mg FICZ CAS: 172922-91-7 FICZ is a potent aryl hydrocarbon receptor (AhR) agonist with a Kd of 70 pM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9660