Learn More
Medchemexpress LLC HY-12451 1mg Medchemexpress, FICZ CAS:172922-91-7 Purity:>98%
SDP
Catalog No. 502028879
Encompass_Preferred
Medchemexpress, HY-12451 1mg FICZ CAS:172922-91-7 FICZ is a potent aryl hydrocarbon receptor (AhR) agonist with a Kd of 70 pM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8879 Supplier Medchemexpress LLC Supplier No. HY124511MG
Medchemexpress, HY-12451 1mg FICZ CAS:172922-91-7 FICZ is a potent aryl hydrocarbon receptor (AhR) agonist with a Kd of 70 pM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.