Learn More
Medchemexpress LLC FIDAS-5 | 1391934-98-7 | 98.5% | 261.72 | 50 MG
SDP
Catalog No. 5000351239
Encompass_Preferred
FIDAS-5 is a potent and orally active methionine S-adenosyltransferase 2A (MAT2A) inhibitor with an IC50 of 2.1 μM. It effectively competes against S-adenosylmethionine (SAM) for MAT2A binding and exhibits anticancer activities.
  • Potent and orally active MAT2A inhibitor
  • Effectively competes against S-adenosylmethionine (SAM)
  • Exhibits anticancer activities
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Compound is unstable in solutions, freshly prepared is recommended
Catalog No. 50-003-51239 Supplier Medchemexpress LLC Supplier No. HY13614450MG
FIDAS-5 is a potent and orally active methionine S-adenosyltransferase 2A (MAT2A) inhibitor with an IC50 of 2.1 μM. It effectively competes against S-adenosylmethionine (SAM) for MAT2A binding and exhibits anticancer activities.
  • Potent and orally active MAT2A inhibitor
  • Effectively competes against S-adenosylmethionine (SAM)
  • Exhibits anticancer activities
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Compound is unstable in solutions, freshly prepared is recommended