Learn More
Medchemexpress LLC HY-136144 5mg Medchemexpress, FIDAS-5 CAS:1391934-98-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1361445MG
Encompass_Preferred
Medchemexpress, HY-136144 5mg FIDAS-5 CAS:1391934-98-7 FIDAS-5 is a potent and orally active methionine S-adenosyltransferase 2A (MAT2A) inhibitor with an IC50 of 2.1 μM. FIDAS-5 effectively competes against S-adenosylmethionine (SAM) for MAT2A binding. FIDAS-5 has anticancer activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0398
 Restricted Item