Learn More
Medchemexpress LLC HY-17580 5mg Medchemexpress, Fidaxomicin CAS:873857-62-6 Purity:>98%
SDP
Catalog No. 502030794
Encompass_Preferred
Medchemexpress, HY-17580 5mg Fidaxomicin CAS:873857-62-6 Fidaxomicin (OPT-80), a macrocyclic RNA polymerase inhibitor, has a narrow spectrum of activity. Fidaxomicin selectively eradicates pathogenic Clostridium difficile with minimal disruption to the multiple species of bacteria that make up the normal, healthy intestinal flora. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0794 Supplier Medchemexpress LLC Supplier No. HY175805MG
Medchemexpress, HY-17580 5mg Fidaxomicin CAS:873857-62-6 Fidaxomicin (OPT-80), a macrocyclic RNA polymerase inhibitor, has a narrow spectrum of activity. Fidaxomicin selectively eradicates pathogenic Clostridium difficile with minimal disruption to the multiple species of bacteria that make up the normal, healthy intestinal flora. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.