Learn More
Medchemexpress LLC Fidaxomicin | 873857-62-6 | MFCD27976367 | 99.9% | 1058.04 | C52H74Cl2O18 | 10 MG
SDP
Catalog No. 5000298528
Encompass_Preferred
Fidaxomicin analytical standard supplied as a measured 10 MG quantity for research and analytical use. It is a macrocyclic antibiotic reference material intended for assay development, method validation, and qualitative or quantitative analyses by HPLC, GC, and MS. The material is provided at high purity suitable for laboratory analytical workflows.
  • High purity suitable for quantitative analysis (99.9%).
  • Intended for HPLC, GC, and MS method development and validation.
  • Provided as a measured 10 MG quantity for analytical use.
  • Useful as a reference standard in qualitative and quantitative assays.
  • Includes CAS number 873857-62-6 for unambiguous identification.
Catalog No. 50-002-98528 Supplier Medchemexpress LLC Supplier No. HY17580R10MG
Fidaxomicin analytical standard supplied as a measured 10 MG quantity for research and analytical use. It is a macrocyclic antibiotic reference material intended for assay development, method validation, and qualitative or quantitative analyses by HPLC, GC, and MS. The material is provided at high purity suitable for laboratory analytical workflows.
  • High purity suitable for quantitative analysis (99.9%).
  • Intended for HPLC, GC, and MS method development and validation.
  • Provided as a measured 10 MG quantity for analytical use.
  • Useful as a reference standard in qualitative and quantitative assays.
  • Includes CAS number 873857-62-6 for unambiguous identification.