Learn More
Medchemexpress LLC Fidaxomicin-d7 | 2143934-06-7 | 97.2% | 1065.08 | 100 UG
SDP
Catalog No. 5000387473
Encompass_Preferred
Fidaxomicin-d7 (OPT-80-D7) is the deuterium labeled Fidaxomicin. Fidaxomicin (OPT-80), a macrocyclic RNA polymerase inhibitor, has a narrow spectrum of activity. Fidaxomicin selectively eradicates pathogenic *Clostridium difficile* with minimal disruption to the multiple species of bacteria that make up the normal, healthy intestinal flora.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-87473 Supplier Medchemexpress LLC Supplier No. HY17580S100UG
Fidaxomicin-d7 (OPT-80-D7) is the deuterium labeled Fidaxomicin. Fidaxomicin (OPT-80), a macrocyclic RNA polymerase inhibitor, has a narrow spectrum of activity. Fidaxomicin selectively eradicates pathogenic *Clostridium difficile* with minimal disruption to the multiple species of bacteria that make up the normal, healthy intestinal flora.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.