Learn More
Cayman Chemical Fidaxomycin, Purity: ≥95%, CAS: 873857-62-6, 50 mg
SDP
Catalog No. 502971262
Encompass_Preferred
A natural macrocyclic antibiotic that inhibits RNA polymerase with selectivity for Gram-positive bacteria over Gram-negative bacteria (IC50s = 0.4 and 6 μM, respectively); has potent antibacterial activity against most Gram-positive bacteria and effectively targets the Gram-positive C. difficile (MIC = 12 ng/ml)
Catalog No. 50-297-1262 Supplier Cayman Chemical Supplier No. 1550350 MG
A natural macrocyclic antibiotic that inhibits RNA polymerase with selectivity for Gram-positive bacteria over Gram-negative bacteria (IC50s = 0.4 and 6 μM, respectively); has potent antibacterial activity against most Gram-positive bacteria and effectively targets the Gram-positive C. difficile (MIC = 12 ng/ml)