Learn More
TARGETMOL CHEMICALS INC FIDEXABAN 5MG
SDP
Catalog No. 502580628
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Fidexaban (CI1031) is a novel potent selective and orally active factor Xa inhibitor that has demonstrated antithrombotic activity in a variety of assayspurity: 99%
Catalog No. 50-258-0628 Supplier TARGETMOL CHEMICALS INC Supplier No. T273185MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Fidexaban (CI1031) is a novel potent selective and orally active factor Xa inhibitor that has demonstrated antithrombotic activity in a variety of assayspurity: 99%