Learn More
Hach Company Field Power Pack for Portable Ultraviolet Lamps 2670800 & 2671000
Catalog No. 501996511 Shop All Hach Company Products
Encompass
Shortwave (254 nm) UV lamps for field or laboratory use. Either lamp can be used inthe lab with power supply while Portable Field Pack can only be used with the1.75" lamp. Portable Field Pack requires a 9-Volt battery (not included) and has abelt clip for easy traveling. Both lamps are physically identical except for bulb and cord length (these are shorter in the 1.75" version). NOTE: Not for use in microbiological applications. UV Lamp contains mercury. Dispose according to state, local or federal regulations.
Catalog No. 50-199-6511 Supplier Hach Company Supplier No. 2670600
Shortwave (254 nm) UV lamps for field or laboratory use. Either lamp can be used inthe lab with power supply while Portable Field Pack can only be used with the1.75" lamp. Portable Field Pack requires a 9-Volt battery (not included) and has abelt clip for easy traveling. Both lamps are physically identical except for bulb and cord length (these are shorter in the 1.75" version). NOTE: Not for use in microbiological applications. UV Lamp contains mercury. Dispose according to state, local or federal regulations.