Learn More
Brady™ Field Wire Input Adapter, Black
Catalog No. 17823774 Shop All Brady Corporation Products
Change view
Click to view available options
Display:
LED
Catalog No. Display
17-823-774 LED
1 options
Catalog No. 17-823-774 Supplier Brady™ Supplier No. 178983
Add to Cart
Add to Cart
Connect external control systems to any Brady LED projector without cutting or damaging the factory wiring.
  • PLC-ready integration: Provides seamless connection to Programmable Logic Controllers (PLCs) and other automated systems.
  • Tool-free: Convenient screw terminals enable fast, secure connections.
  • Fully compatible: For use with any Brady LED projector.
  • Voltage: Supports DC input signals up to 10 V.
Specifications
Product Type Cable
Display LED