Learn More
Medchemexpress LLC HY-18603 5mg Medchemexpress, FIIN-3 CAS:1637735-84-2 Purity:>98%
SDP
Catalog No. 502082054
Encompass_Preferred
Medchemexpress, HY-18603 5mg FIIN-3 CAS:1637735-84-2 FIIN-3 is an irreversible inhibitor of FGFR with an IC50 of 13.1, 21, 31.4, and 35.3 nM for FGFR1, FGFR2, FGFR3 and FGFR4, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2054 Supplier Medchemexpress LLC Supplier No. HY186035MG
Medchemexpress, HY-18603 5mg FIIN-3 CAS:1637735-84-2 FIIN-3 is an irreversible inhibitor of FGFR with an IC50 of 13.1, 21, 31.4, and 35.3 nM for FGFR1, FGFR2, FGFR3 and FGFR4, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.