Learn More
Enzo Life Sciences Filaggrin polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS1810100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 24-39 in the partial sequence of filaggrin. Species reactivity: Human, Mouse Primate. Applications: IF, IHC, WB. Purity: Affinity purified. Formulation: Liquid. In PBS containing 0.03% thimerosal.
Catalog No. 50-201-2733