Learn More
Sutter Instruments FILAMENT 3.0 WIDE
SDP
Catalog No. NC9465664
Encompass_Preferred
Trough Filaments , 3.0mm wide trough filament, General purpose filament and are recommended for patch pipettes produced from either standard or thin wall glass
Catalog No. NC9465664 Supplier Sutter Instruments Supplier No. FGFT330B
Trough Filaments , 3.0mm wide trough filament, General purpose filament and are recommended for patch pipettes produced from either standard or thin wall glass